Search
Search

Smarcc1| Gene Symbol: Smarcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834968.2 | XP_004835025.1 | 16 - 17 | 1582 | 63 | |
| XM_021256732.1 | XP_021112391.1 | 16 - 17 | 1582 | 63 | ||
| XM_021256736.1 | XP_021112395.1 | 16 - 17 | 1540 | 63 | ||
| XM_021256739.1 | XP_021112398.1 | 16 - 17 | 1582 | 63 | ||
Set Membership: |
|