| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000507179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000507179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000507179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000508416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
ENST00000508416.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000818.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000818.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000818.hg.1 |
TC01000057.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000822.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000822.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000822.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000822.hg.1 |
TC01000057.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000817.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000817.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000817.hg.1 |
TC01000057.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000821.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000821.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000821.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01000821.hg.1 |
TC01000057.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000507179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000507179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000507179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000508416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
ENST00000508416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006565.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100145341.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145341.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100145341.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145341.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100145341.hg.1 |
TC0100006565.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100145343.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145343.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100145343.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145343.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100145343.hg.1 |
TC0100006565.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100145347.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145347.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145347.hg.1 |
ENST00000508416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145347.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100145347.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145347.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145347.hg.1 |
ENST00000508416.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145347.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100145347.hg.1 |
TC0100006565.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100145348.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145348.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145348.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100145348.hg.1 |
ENST00000378536.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145348.hg.1 |
ENST00000478223.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100145348.hg.1 |
NM_003036.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100145348.hg.1 |
TC0100006565.hg.1 |
Clariom D TC |