| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002974.hg.1 |
ENST00000261650.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002974.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002974.hg.1 |
TC17000207.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002977.hg.1 |
ENST00000261650.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002977.hg.1 |
ENST00000399138.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002977.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002977.hg.1 |
TC17000207.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002978.hg.1 |
ENST00000261650.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002978.hg.1 |
ENST00000399138.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002978.hg.1 |
ENST00000500385.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002978.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002978.hg.1 |
TC17000207.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002979.hg.1 |
ENST00000261650.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002979.hg.1 |
ENST00000399138.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002979.hg.1 |
ENST00000500385.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002979.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002979.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002979.hg.1 |
TC17000207.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002976.hg.1 |
ENST00000261650.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002976.hg.1 |
ENST00000399138.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002976.hg.1 |
ENST00000500385.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002976.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002976.hg.1 |
TC17000207.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009966.hg.1 |
ENST00000577847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009966.hg.1 |
ENST00000577847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009966.hg.1 |
ENST00000577847.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009966.hg.1 |
ENST00000583062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009966.hg.1 |
ENST00000583062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009966.hg.1 |
ENST00000583062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700009966.hg.1 |
ENST00000583062.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002975.hg.1 |
ENST00000261650.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002975.hg.1 |
ENST00000500385.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002975.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17002975.hg.1 |
TC17000207.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1700150188.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150188.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150188.hg.1 |
TC1700007102.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700150188.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150188.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700009966.hg.1 |
ENST00000577847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009966.hg.1 |
ENST00000577847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009966.hg.1 |
ENST00000577847.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009966.hg.1 |
ENST00000583062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009966.hg.1 |
ENST00000583062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009966.hg.1 |
ENST00000583062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700009966.hg.1 |
ENST00000583062.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150189.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150189.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150189.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150189.hg.1 |
TC1700007102.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700150189.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150189.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700150189.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000399138.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000490106.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
ENST00000541285.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700007102.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150185.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150185.hg.1 |
TC1700007102.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700150185.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150186.hg.1 |
NM_017758.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700150186.hg.1 |
TC1700007102.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700150186.hg.1 |
NM_017758.3 |
Refseq |