| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006303.hg.1 |
ENST00000537169.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006303.hg.1 |
NM_177948.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X006303.hg.1 |
TC0X000481.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
ENST00000467808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
ENST00000477980.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
ENST00000491568.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
ENST00000537169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
NM_177948.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
ENST00000467808.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
ENST00000477980.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
ENST00000491568.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
ENST00000537169.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
NM_177948.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00154141.hg.1 |
TC0X00007951.hg.1 |
Clariom D TC |