| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000509913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000509913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000509913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
ENST00000509913.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024221.hg.1 |
ENST00000231512.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024221.hg.1 |
ENST00000451255.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024221.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024221.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024221.hg.1 |
TC05001784.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024223.hg.1 |
ENST00000231512.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024223.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024223.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05024223.hg.1 |
TC05001784.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500183359.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183359.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183359.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183359.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183359.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500183359.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500183359.hg.1 |
TC0500012057.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000231512.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000507191.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000509913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000509913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000509913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
ENST00000509913.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500012057.hg.1 |
NM_020199.2 |
Refseq |