| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010558.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010558.hg.1 |
ENST00000511601.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010558.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04010558.hg.1 |
TC04000768.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400009029.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161915.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161915.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161915.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161915.hg.1 |
ENST00000260008.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161915.hg.1 |
ENST00000511601.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400161915.hg.1 |
NM_033393.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400161915.hg.1 |
TC0400009029.hg.1 |
Clariom D TC |