| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001894.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001894.hg.1 |
ENST00000315901.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001894.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01001894.hg.1 |
TC01000124.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0100147067.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100147067.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100147067.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100147067.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100147067.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100147067.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100147067.hg.1 |
TC0100006805.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000294435.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
ENST00000315901.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100006805.hg.1 |
NM_052960.2 |
Refseq |