| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000543316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000543316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000543316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000543316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016576.hg.1 |
ENST00000261167.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016576.hg.1 |
ENST00000537574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016576.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016576.hg.1 |
TC12001273.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016577.hg.1 |
ENST00000261167.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016577.hg.1 |
ENST00000537574.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016577.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12016577.hg.1 |
TC12001273.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200175632.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175632.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175632.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200175632.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175632.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175632.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200175632.hg.1 |
TC1200010016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1200175631.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175631.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175631.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200175631.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175631.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200175631.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200175631.hg.1 |
TC1200010016.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000261167.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535328.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000535638.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000543316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000543316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000543316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000543316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
ENST00000544764.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1200010016.hg.1 |
NM_016312.2 |
Refseq |