| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000471030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000471030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000471030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000471030.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000486022.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000486022.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000486022.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
ENST00000486022.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000312849.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000439068.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000455302.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000471030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000471030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000471030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000471030.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000486022.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000486022.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000486022.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
ENST00000486022.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_001134468.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013442.hg.1 |
NM_182597.2 |
Refseq |