| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000465320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000465320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000465320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000465320.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000481329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000481329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000481329.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000491641.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000491641.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000491641.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07017499.hg.1 |
ENST00000350796.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07017499.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07017499.hg.1 |
TC07001127.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000461891.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000465320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000465320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000465320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000465320.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000466543.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000481329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000481329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000481329.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000482460.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000488964.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000491641.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000491641.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000491641.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
ENST00000495176.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700010189.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700172494.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700172494.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700172494.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700172494.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700172494.hg.1 |
ENST00000350796.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700172494.hg.1 |
ENST00000396741.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700172494.hg.1 |
ENST00000478541.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700172494.hg.1 |
NM_004227.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700172494.hg.1 |
TC0700010189.hg.1 |
Clariom D TC |