| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03017336.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03017336.hg.1 |
ENST00000426955.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03017336.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03017336.hg.1 |
TC03000996.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03017337.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03017337.hg.1 |
ENST00000426955.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03017337.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03017337.hg.1 |
TC03000996.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300170092.hg.1 |
TC0300009696.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000273794.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000426955.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000461141.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000474580.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000493493.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
ENST00000497229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300009696.hg.1 |
NM_138345.2 |
Refseq |