Search
Search

EPHA7| Gene Symbol: EPHA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001288629.1 | NP_001275558.1 | 12 - 13 | 2392 | 93 | |
| NM_004440.3 | NP_004431.1 | 12 - 13 | 2407 | 93 | ||
| XM_005248669.2 | XP_005248726.1 | 12 - 13 | 2452 | 93 | ||
| XM_017010365.1 | XP_016865854.1 | 12 - 13 | 2437 | 93 | ||
| GenBank mRNA | AB209269.1 | - | 12 - 13 | 2379 | 93 | |
| AK313529.1 | - | 12 - 13 | 2379 | 93 | ||
| BC126125.1 | - | 12 - 13 | 2281 | 8991841 | 93 | |
| BC126151.1 | - | 12 - 13 | 2281 | 8991867 | 93 | |
| BC143857.1 | - | 12 - 13 | 2266 | 9052373 | 93 | |
| BC143858.1 | - | 12 - 13 | 2269 | 9052374 | 93 | |
| L36642.1 | - | 12 - 13 | 2351 | 93 | ||
Set Membership: |
|