| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
ENST00000329411.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
ENST00000335847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
ENST00000355791.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
ENST00000507853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
ENST00000536021.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021590.hg.1 |
TC06001788.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021593.hg.1 |
ENST00000335847.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021593.hg.1 |
ENST00000507853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021593.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021593.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06021593.hg.1 |
TC06001788.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600012006.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182629.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182629.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182629.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182629.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182629.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182629.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182629.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182629.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182629.hg.1 |
TC0600012006.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
ENST00000329411.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
ENST00000335847.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
ENST00000505118.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
ENST00000507853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
NM_001131.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
NM_001205220.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
NM_170609.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600182626.hg.1 |
TC0600012006.hg.1 |
Clariom D TC |