| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17025397.hg.1 |
ENST00000254816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17025397.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17025397.hg.1 |
TC17001887.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17025398.hg.1 |
ENST00000254816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17025398.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17025398.hg.1 |
TC17001887.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17025395.hg.1 |
ENST00000254816.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17025395.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17025395.hg.1 |
TC17001887.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000585333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000585333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000585333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000585333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000586495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000586495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000586495.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000593089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000593089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
ENST00000593089.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700195808.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195808.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700195808.hg.1 |
TC1700011774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700195808.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195808.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700195807.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195807.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700195807.hg.1 |
TC1700011774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700195807.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195807.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000585333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000585333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000585333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000585333.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000586495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000586495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000586495.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000592942.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000593089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000593089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
ENST00000593089.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700011774.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700195804.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195804.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195804.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195804.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700195804.hg.1 |
TC1700011774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700195804.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195804.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195804.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195804.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700195805.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195805.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195805.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195805.hg.1 |
NM_033452.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1700195805.hg.1 |
TC1700011774.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1700195805.hg.1 |
ENST00000254816.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195805.hg.1 |
ENST00000587339.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195805.hg.1 |
ENST00000587774.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1700195805.hg.1 |
NM_033452.2 |
Refseq |