Search
Search

ATP9A| Gene Symbol: ATP9A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006045.2 | NP_006036.1 | 26 - 27 | 2847 | 108 | |
| XM_011528481.2 | XP_011526783.1 | 26 - 27 | 2875 | 108 | ||
| GenBank mRNA | AB014511.1 | - | 21 - 22 | 2401 | 108 | |
| AK172802.1 | - | 24 - 25 | 2536 | 108 | ||
| AK299064.1 | - | 26 - 27 | 2776 | 108 | ||
| BC110591.1 | - | 22 - 23 | 2339 | 108 | ||
| BC110592.1 | - | 22 - 23 | 2339 | 108 | ||
Set Membership: |
|