Search
Search
| Gene Symbol: TSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015362.1 | 6 - 7 | 2080 | 158 | ||
| GenBank mRNA | AY314745.1 | - | 6 - 7 | 2080 | 158 | |
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | EPH receptor A7 | EPHA7 |
EHK-3, EHK3, EK11, HEK11 |
-324.5kb | Anti-sense | Chr. 6 - 93240020 - 93419687 on NCBI Build GRCh38 |
| Downstream | MANEA antisense RNA 1 (head to head) | MANEA-AS1 | 1.8mb | Anti-sense | Chr. 6 - 95560096 - 95577451 on NCBI Build GRCh38 |
Set Membership: |
|