| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000317578.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560160.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000560168.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
ENST00000622857.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900010953.hg.1 |
NM_175875.4 |
Refseq |