Search
Search
| Gene Symbol: UPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006760.3 | NP_006751.1 | 1 - 2 | 126 | 80 | |
| GenBank mRNA | AY455312.1 | - | 1 - 2 | 73 | 80 | |
| BC113900.1 | - | 1 - 2 | 104 | 40029623 | 80 | |
| Y13645.1 | - | 1 - 2 | 111 | 80 | ||
Set Membership: |
|