| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153166.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153166.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153166.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153166.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153166.hg.1 |
TC1400007585.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400153167.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153167.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153167.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153167.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153167.hg.1 |
TC1400007585.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400153168.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153168.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153168.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153168.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153168.hg.1 |
TC1400007585.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400007585.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153170.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153170.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153170.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153170.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153170.hg.1 |
TC1400007585.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1400153169.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153169.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153169.hg.1 |
ENST00000553682.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400153169.hg.1 |
NR_125769.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400153169.hg.1 |
TC1400007585.hg.1 |
Clariom D TC |