| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009927.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009927.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009927.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009927.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009927.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009927.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009927.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009927.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009927.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500170069.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500170069.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500170069.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500170069.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500170069.hg.1 |
TC0500009927.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500170068.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500170068.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500170068.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500170068.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500170068.hg.1 |
TC0500009927.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500170070.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500170070.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500170070.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500170070.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500170070.hg.1 |
TC0500009927.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500170071.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500170071.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500170071.hg.1 |
ENST00000508823.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500170071.hg.1 |
NR_104618.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500170071.hg.1 |
TC0500009927.hg.1 |
Clariom D TC |