| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000506008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000506008.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000357504.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000456874.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000503528.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000506008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000506008.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
ENST00000507598.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001136536.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007225.hg.1 |
NM_001170700.2 |
Refseq |