| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400008309.hg.1 |
ENST00000606234.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
ENST00000500179.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008310.hg.1 |
NR_125926.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400008309.hg.1 |
ENST00000606234.1 |
Ensembl |