| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000562637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000562637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000562637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000562637.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000564848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000564848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000564848.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000565074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000565074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000565074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000360639.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000394987.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000562637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000562637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000562637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000562637.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000563905.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000564848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000564848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000564848.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000565074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000565074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000565074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
ENST00000567617.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007874.hg.1 |
NM_015492.4 |
Refseq |