Search
Search

SYCP2| Gene Symbol: SYCP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005569500.2 | XP_005569557.1 | 43 - 44 | 4565 | 72 | |
| XM_005569501.2 | XP_005569558.1 | 43 - 44 | 4637 | 72 | ||
| XM_005569503.2 | XP_005569560.1 | 36 - 37 | 3947 | 72 | ||
| XM_015430024.1 | XP_015285510.1 | 42 - 43 | 4720 | 72 | ||
Set Membership: |
|