Search
Search

SIK3| Gene Symbol: SIK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579733.2 | XP_005579790.1 | 4 - 5 | 554 | 151 | |
| XM_005579736.1 | XP_005579793.1 | 4 - 5 | 554 | 151 | ||
| XM_005579737.2 | XP_005579794.1 | 4 - 5 | 1664 | 151 | ||
| XM_015435751.1 | XP_015291237.1 | 4 - 5 | 554 | 151 | ||
| XM_015435752.1 | XP_015291238.1 | 4 - 5 | 743 | 151 | ||
| XM_015435753.1 | XP_015291239.1 | 2 - 3 | 1516 | 151 | ||
| XM_015435754.1 | XP_015291240.1 | 3 - 4 | 1600 | 151 | ||
Set Membership: |
|