Search
Search

BOC| Gene Symbol: BOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548136.2 | XP_005548193.1 | 16 - 17 | 3005 | 60 | |
| XM_005548140.2 | XP_005548197.1 | 14 - 15 | 2853 | 60 | ||
| XM_015446412.1 | XP_015301898.1 | 16 - 17 | 3147 | 60 | ||
| XM_015446413.1 | XP_015301899.1 | 15 - 16 | 3087 | 60 | ||
| XM_015446415.1 | XP_015301901.1 | 16 - 17 | 3332 | 60 | ||
| XM_015446416.1 | XP_015301902.1 | 15 - 16 | 2945 | 60 | ||
| XM_015446417.1 | XP_015301903.1 | 16 - 17 | 3644 | 60 | ||
| XM_015446418.1 | XP_015301904.1 | 15 - 16 | 2941 | 60 | ||
| XM_015446419.1 | XP_015301905.1 | 16 - 17 | 3190 | 60 | ||
| XM_015446420.1 | XP_015301906.1 | 15 - 16 | 3442 | 60 | ||
| XM_015446421.1 | XP_015301907.1 | 14 - 15 | 3130 | 60 | ||
| XM_015446422.1 | XP_015301908.1 | 15 - 16 | 3438 | 60 | ||
| XM_015446423.1 | XP_015301909.1 | 16 - 17 | 3144 | 60 | ||
| XM_015446424.1 | XP_015301910.1 | 15 - 16 | 2938 | 60 | ||
| XM_015446425.1 | XP_015301911.1 | 16 - 17 | 3147 | 60 | ||
Set Membership: |
|