Search
Search

RC3H2| Gene Symbol: RC3H2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005580835.2 | XP_005580892.1 | 18 - 19 | 3355 | 128 | |
| XM_005580836.2 | XP_005580893.1 | 18 - 19 | 3330 | 128 | ||
| XM_005580837.2 | XP_005580894.1 | 17 - 18 | 3148 | 128 | ||
| XM_005580838.2 | XP_005580895.1 | 17 - 18 | 3124 | 128 | ||
| XM_005580839.2 | XP_005580896.1 | 18 - 19 | 3295 | 128 | ||
| XM_005580840.2 | XP_005580897.1 | 17 - 18 | 2986 | 128 | ||
| XM_015436440.1 | XP_015291926.1 | 17 - 18 | 4062 | 128 | ||
| XM_015436441.1 | XP_015291927.1 | 15 - 16 | 3883 | 128 | ||
Set Membership: |
|