Search
Search

RASAL2| Gene Symbol: RASAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540082.2 | XP_005540139.1 | 17 - 18 | 4027 | 92 | |
| XM_005540084.2 | XP_005540141.1 | 17 - 18 | 4006 | 92 | ||
| XM_015446138.1 | XP_015301624.1 | 15 - 16 | 6484 | 92 | ||
Set Membership: |
|