Search
Search

SMIM29| Gene Symbol: SMIM29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553263.2 | XP_005553320.1 | 4 - 5 | 807 | 69 | |
| XM_005553264.2 | XP_005553321.1 | 4 - 5 | 699 | 69 | ||
| XM_005553265.2 | XP_005553322.1 | 4 - 5 | 444 | 69 | ||
| XM_005553266.2 | XP_005553323.1 | 4 - 5 | 385 | 69 | ||
| XM_005553267.2 | XP_005553324.1 | 3 - 4 | 216 | 69 | ||
| XM_015449548.1 | XP_015305034.1 | 4 - 5 | 676 | 69 | ||
| XM_015449549.1 | XP_015305035.1 | 4 - 5 | 639 | 69 | ||
| XM_015449550.1 | XP_015305036.1 | 4 - 5 | 626 | 69 | ||
Set Membership: |
|