Search
Search

SIPA1L3| Gene Symbol: SIPA1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589067.2 | XP_005589124.1 | 3 - 4 | 2871 | 61 | |
| XM_005589068.2 | XP_005589125.1 | 3 - 4 | 2466 | 61 | ||
| XM_005589069.2 | XP_005589126.1 | 3 - 4 | 4380 | 61 | ||
| XM_015441297.1 | XP_015296783.1 | 3 - 4 | 2466 | 61 | ||
Set Membership: |
|