Search
Search

SARS2| Gene Symbol: SARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284009.1 | NP_001270938.1 | 9 - 10 | 943 | 83 | |
| XM_005589153.2 | XP_005589210.1 | 10 - 11 | 951 | 83 | ||
| XM_015440934.1 | XP_015296420.1 | 8 - 9 | 915 | 83 | ||
| GenBank mRNA | AB169813.1 | - | 9 - 10 | 943 | 83 | |
Set Membership: |
|