Search
Search

HECTD2| Gene Symbol: HECTD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565939.2 | XP_005565996.1 | 14 - 15 | 1586 | 111 | |
| XM_015456381.1 | XP_015311867.1 | 14 - 15 | 3562 | 111 | ||
| XM_015456382.1 | XP_015311868.1 | 13 - 14 | 1658 | 111 | ||
Set Membership: |
|