Search
Search

DOCK3| Gene Symbol: DOCK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547251.2 | XP_005547308.1 | 54 | 9054 | 137 | |
| XM_005547252.2 | XP_005547309.1 | 54 | 9036 | 137 | ||
| XM_005547253.2 | XP_005547310.1 | 53 | 9028 | 137 | ||
| XM_005547254.2 | XP_005547311.1 | 54 | 9013 | 137 | ||
| XM_005547256.2 | XP_005547313.1 | 53 | 8931 | 137 | ||
| XM_005547258.2 | XP_005547315.1 | 48 | 8364 | 137 | ||
| XM_015445960.1 | XP_015301446.1 | 54 | 8989 | 137 | ||
Set Membership: |
|