Search
Search

SLC6A4| Gene Symbol: SLC6A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287324.1 | NP_001274253.1 | 5 - 6 | 999 | 60 | |
| XM_005583332.2 | XP_005583389.1 | 4 - 5 | 1679 | 60 | ||
| XM_005583333.2 | XP_005583390.1 | 4 - 5 | 1085 | 60 | ||
| XM_005583334.2 | XP_005583391.1 | 4 - 5 | 3670 | 60 | ||
| GenBank mRNA | EF126285.1 | - | 5 - 6 | 999 | 60 | |
Set Membership: |
|