Search
Search

CTNNB1| Gene Symbol: CTNNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319394.1 | NP_001306323.1 | 14 - 15 | 2295 | 72 | |
| XM_005546747.2 | XP_005546804.1 | 13 - 14 | 2344 | 72 | ||
| XM_005546748.2 | XP_005546805.1 | 13 - 14 | 2344 | 72 | ||
| XM_005546749.2 | XP_005546806.1 | 14 - 15 | 2477 | 72 | ||
| XM_015445703.1 | XP_015301189.1 | 13 - 14 | 2364 | 72 | ||
| XM_015445705.1 | XP_015301191.1 | 14 - 15 | 2457 | 72 | ||
| GenBank mRNA | AB179333.1 | - | 14 - 15 | 2295 | 72 | |
| AY650367.1 | - | 11 - 12 | 1875 | 72 | ||
Set Membership: |
|