Search
Search

IDH2| Gene Symbol: IDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284588.1 | NP_001271517.1 | 10 - 11 | 1354 | 59 | |
| XM_015453247.1 | XP_015308733.1 | 8 - 9 | 1176 | 59 | ||
| GenBank mRNA | AB169742.1 | - | 10 - 11 | 1354 | 59 | |
Set Membership: |
|