Search
Search

CNPPD1| Gene Symbol: CNPPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005574343.2 | XP_005574400.1 | 1 - 2 | 988 | 61 | |
| XM_015432832.1 | XP_015288318.1 | 1 - 2 | 985 | 61 | ||
Set Membership: |
|