Search
Search

ABHD12| Gene Symbol: ABHD12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284729.1 | NP_001271658.1 | 6 - 7 | 616 | 85 | |
| XM_005568131.2 | XP_005568188.1 | 6 - 7 | 633 | 85 | ||
| XM_015457647.1 | XP_015313133.1 | 6 - 7 | 633 | 85 | ||
| XM_015457648.1 | XP_015313134.1 | 6 - 7 | 473 | 85 | ||
| GenBank mRNA | AB168958.1 | - | 6 - 7 | 616 | 85 | |
Set Membership: |
|