Search
Search

TAPT1| Gene Symbol: TAPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554528.2 | XP_005554585.1 | 8 - 9 | 1009 | 101 | |
| XM_005554530.2 | XP_005554587.1 | 8 - 9 | 948 | 101 | ||
| XM_015450198.1 | XP_015305684.1 | 7 - 8 | 884 | 101 | ||
Set Membership: |
|