Search
Search

ECD| Gene Symbol: ECD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565531.2 | XP_005565588.1 | 8 - 9 | 1260 | 73 | |
| XM_005565532.1 | XP_005565589.1 | 8 - 9 | 1282 | 73 | ||
| XM_005565534.2 | XP_005565591.1 | 7 - 8 | 1153 | 73 | ||
| XM_015456186.1 | XP_015311672.1 | 7 - 8 | 1194 | 73 | ||
| XM_015456187.1 | XP_015311673.1 | 7 - 8 | 1192 | 73 | ||
| XM_015456188.1 | XP_015311674.1 | 6 - 7 | 1065 | 73 | ||
| XM_015456189.1 | XP_015311675.1 | 6 - 7 | 1064 | 73 | ||
| GenBank mRNA | AB171175.1 | - | 8 - 9 | 1228 | 73 | |
Set Membership: |
|