Search
Search

SFTPB| Gene Symbol: SFTPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005575398.2 | XP_005575455.1 | 1 - 2 | 121 | 92 | |
| XM_005575399.2 | XP_005575456.1 | 1 - 2 | 121 | 92 | ||
| XM_005575400.2 | XP_005575457.1 | 1 - 2 | 121 | 92 | ||
| XM_005575401.2 | XP_005575458.1 | 1 - 2 | 121 | 92 | ||
| XM_005575403.2 | XP_005575460.1 | 1 - 2 | 111 | 92 | ||
Set Membership: |
|