Search
Search

MARVELD2| Gene Symbol: MARVELD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557093.2 | XP_005557150.1 | 4 - 5 | 1712 | 104 | |
| XM_005557094.2 | XP_005557151.1 | 4 - 5 | 1441 | 104 | ||
| XM_005557095.2 | XP_005557152.1 | 4 - 5 | 1516 | 104 | ||
Set Membership: |
|