Search
Search

ZRANB3| Gene Symbol: ZRANB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573008.2 | XP_005573065.2 | 7 - 8 | 969 | 100 | |
| XM_005573009.2 | XP_005573066.1 | 7 - 8 | 1011 | 100 | ||
| XM_005573010.2 | XP_005573067.1 | 7 - 8 | 1023 | 100 | ||
| XM_005573011.2 | XP_005573068.1 | 7 - 8 | 967 | 100 | ||
| XM_005573012.2 | XP_005573069.1 | 7 - 8 | 946 | 100 | ||
| XM_005573013.2 | XP_005573070.1 | 7 - 8 | 971 | 100 | ||
| XM_005573015.2 | XP_005573072.1 | 5 - 6 | 700 | 100 | ||
| XM_005573016.2 | XP_005573073.1 | 6 - 7 | 2177 | 100 | ||
| XM_005573018.2 | XP_005573075.1 | 6 - 7 | 1463 | 100 | ||
| XM_005573019.2 | XP_005573076.1 | 5 - 6 | 768 | 100 | ||
| XM_005573020.2 | XP_005573077.1 | 5 - 6 | 1025 | 100 | ||
| XM_005573022.2 | XP_005573079.2 | 2 - 3 | 1014 | 100 | ||
| XM_015432030.1 | XP_015287516.1 | 7 - 8 | 1067 | 100 | ||
| XM_015432031.1 | XP_015287517.1 | 7 - 8 | 2309 | 100 | ||
| XM_015432032.1 | XP_015287518.1 | 3 - 4 | 374 | 100 | ||
Set Membership: |
|