Search
Search

ATG4D| Gene Symbol: ATG4D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587954.2 | XP_005588011.1 | 4 - 5 | 970 | 130 | |
| XM_005587955.2 | XP_005588012.1 | 4 - 5 | 1041 | 130 | ||
| GenBank mRNA | AB168133.1 | - | 4 - 5 | 677 | 130 | |
Set Membership: |
|