Search
Search

TSPOAP1| Gene Symbol: TSPOAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583830.2 | XP_005583887.1 | 25 - 26 | 9753 | 136 | |
| XM_005583832.2 | XP_005583889.1 | 24 - 25 | 9572 | 136 | ||
| XM_015438351.1 | XP_015293837.1 | 24 - 25 | 9840 | 136 | ||
| XM_015438352.1 | XP_015293838.1 | 23 - 24 | 9819 | 136 | ||
| XM_015438354.1 | XP_015293840.1 | 24 - 25 | 9840 | 136 | ||
| XM_015438356.1 | XP_015293842.1 | 25 - 26 | 9754 | 136 | ||
| XM_015438357.1 | XP_015293843.1 | 24 - 25 | 9839 | 136 | ||
| XM_015438358.1 | XP_015293844.1 | 24 - 25 | 9839 | 136 | ||
| XM_015438359.1 | XP_015293845.1 | 23 - 24 | 9659 | 136 | ||
| XM_015438360.1 | XP_015293846.1 | 24 - 25 | 9841 | 136 | ||
| XM_015438361.1 | XP_015293847.1 | 24 - 25 | 9841 | 136 | ||
| XM_015438362.1 | XP_015293848.1 | 23 - 24 | 9001 | 136 | ||
| XM_015438364.1 | XP_015293850.1 | 24 - 25 | 8914 | 136 | ||
| XM_015438366.1 | XP_015293852.1 | 20 - 21 | 4424 | 136 | ||
| XM_015438367.1 | XP_015293853.1 | 23 - 24 | 8733 | 136 | ||
| XM_015438368.1 | XP_015293854.1 | 20 - 21 | 4406 | 136 | ||
Set Membership: |
|