Search
Search

GPC6| Gene Symbol: GPC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586087.2 | XP_005586144.1 | 6 - 7 | 1785 | 72 | |
| XM_015439417.1 | XP_015294903.1 | 7 - 8 | 1215 | 72 | ||
| XM_015439418.1 | XP_015294904.1 | 7 - 8 | 3493 | 72 | ||
| XM_015439419.1 | XP_015294905.1 | 8 - 9 | 4573 | 72 | ||
| XM_015439420.1 | XP_015294906.1 | 7 - 8 | 1859 | 72 | ||
| XM_015439421.1 | XP_015294907.1 | 7 - 8 | 1188 | 72 | ||
Set Membership: |
|