Search
Search

RNF123| Gene Symbol: RNF123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547144.2 | XP_005547201.1 | 6 - 7 | 513 | 72 | |
| XM_005547145.2 | XP_005547202.1 | 6 - 7 | 513 | 72 | ||
| XM_005547146.2 | XP_005547203.1 | 4 - 5 | 303 | 72 | ||
| XM_015445910.1 | XP_015301396.1 | 5 - 6 | 1712 | 72 | ||
| XM_015445911.1 | XP_015301397.1 | 6 - 7 | 449 | 72 | ||
Set Membership: |
|