Search
Search

KLRF1| Gene Symbol: KLRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285324.1 | NP_001272253.1 | 4 - 5 | 480 | 109 | |
| XM_015430177.1 | XP_015285663.1 | 3 - 4 | 1002 | 109 | ||
| GenBank mRNA | AJ426430.1 | - | 4 - 5 | 480 | 109 | |
Set Membership: |
|