Search
Search

LOC101867327| Gene Symbol: LOC101867327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283528.1 | NP_001270457.1 | 1 - 2 | 271 | 101 | |
| GenBank mRNA | AB168377.1 | - | 1 - 2 | 271 | 101 | |
Set Membership: |
|