Search
Search

CR1| Gene Symbol: CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015449302.1 | XP_015304788.1 | 3 - 4 | 2050 | 105 | |
| XM_015449307.1 | XP_015304793.1 | 3 - 4 | 2050 | 105 | ||
| XM_015449309.1 | XP_015304795.1 | 2 - 3 | 1908 | 105 | ||
| XM_015449313.1 | XP_015304799.1 | 3 - 4 | 1317 | 105 | ||
| XM_015449318.1 | XP_015304804.1 | 3 - 4 | 2056 | 105 | ||
| XM_015449320.1 | XP_015304806.1 | 3 - 4 | 1318 | 105 | ||
| XM_015449323.1 | XP_015304809.1 | 3 - 4 | 2051 | 105 | ||
Set Membership: |
|